Learn More
MilliporeSigma™ Supelco™ SPME Inlet Guide
Catalog No. 111004869 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Aluminum
Catalog No. Material
11-100-4869 Aluminum
1 options
Catalog No. 11-100-4869 Supplier MilliporeSigma™ Supelco™ Supplier No. 57356U
Add to Cart
Add to Cart
Secures the SPME fiber holder in the injection port during the thermal desorption process. Interchangeable among Merlin Microseal sealing system and most Varian and Agilent chromatographs.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Food and Beverages
For Use With (Equipment) Merlin Microseal Sealing System and Most Varian and Agilent Chromatographs
Type SPE Accessory
Material Aluminum