Learn More
MilliporeSigma™ Supelco™ SPME LC C18 Tips
Catalog No. 111009194 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
96 Tips/Tray
Catalog No. Quantity
11-100-9194 96 Tips/Tray
1 options
Catalog No. 11-100-9194 Supplier MilliporeSigma™ Supelco™ Supplier No. 57234U
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Low cost, micro-sample prep devices consisting of a coated microfiber held in place in a micropipette tip making it compatible with both multi-channel pipettors and robotic liquid handling systems. The tips are sold in a 96 well tray configuration.
TRUSTED_SUSTAINABILITY
Specifications
Column Type SPE
Description Fiber L: 1 cm
For Use With (Application) Food and Beverages
For Use With (Equipment) Pipettor or Robotic Liquid Handling Systems
Type SPME Tips
Quantity 96 Tips/Tray