Learn More
MilliporeSigma™ Supelco™ SPME Portable Field Sampler
Catalog No. 111003760 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Carboxen/Polydimethylsiloxane Coating
Polydimethylsiloxane Coating
Polydimethylsiloxane/Divinylbenzene Coating
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
11-100-3760 Polydimethylsiloxane/Divinylbenzene Coating
11-100-3712 Carboxen/Polydimethylsiloxane Coating
11-100-3711 Polydimethylsiloxane Coating
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-100-3760 Supplier MilliporeSigma™ Supelco™ Supplier No. 57359U
Add to Cart
Add to Cart
Efficient and economical way of extracting and transporting volatile and semivolatile compounds from field samples.
Concentrate and Store Analytes from Water; Sample Indoor Air - The SPME portable field sampler is an efficient and economical way of extracting and transporting volatile and semivolatile compounds from field samples. Extracted compounds storage losses for pesticides extracted and stored using a portable field sampler were significantly lower than losses from stored whole water samples. The sampler can be reused 50-100 times, and is disposed of when the fiber is no longer usable.The portable field sampler also efficiently detects organic compounds in air. In our studies, the sampler allowed us to monitor typical HPLC and GC solvents at ppb levels in laboratory air. Four fibers are available: a polydimethylsiloxane(PDMS)/Carboxen fiber for trace levels of volatiles, a general purpose PDMS fiber, a PDMS/DVB fiber for semi-volatiles and larger volatiles, and a Carbowax/DVB fiber for polar semivolatiles.Five slots in the needle guide/depth gauge control the depth of needle insertion into a sample container, or into the injection port during fiber desorption. Assemblies contain 24 gauge needles. 23 gauge needles and other coatings may be available as customs.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Food and Beverages
Type SPME Fibers and Accessories
Material Polydimethylsiloxane/Divinylbenzene Coating
Quantity 2/Pk.