Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SPME Sampling Stand

Catalog No. 111016229 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
For use with 15 mL vials
For use with 4 mL vials
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-101-6229 For use with 4 mL vials
11-100-3610 For use with 15 mL vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-6229 Supplier MilliporeSigma™ Supelco™ Supplier No. 57333U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Holds eight vials while supporting the SPME syringe for consistent fiber immersion depth. Cat. No. 57333-U accommodates 4 mL vials only; Cat. No. 57357-U accommodates 15 mL vials. Order the 15 mL vial puck (Cat. No. 57358-U) as a replacement for the 15 mL unit, or to use 15 mL vials with the 4 mL unit. Not for use with automated/HPLC fiber holders.

TRUSTED_SUSTAINABILITY

Specifications

Description For use with 4 mL vials
For Use With (Application) Food and Beverages
Type SPME Fibers and Accessories
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.