Learn More
Med Vet International McKesson Sterile Nonwoven Sponge, 4 x 4 Inch, 16-2436, 1200/Case
Catalog No. 503045503
Encompass_Preferred
Nonwoven Sponge McKesson Nonwoven / Rayon / Cellulose 4 X 4 Inch Square Sterile Nonwoven, 4 x 4 inch sterile sponges are ideal for moderate to high exuding wounds They have a rayon and cellulose absorbent core The nonwoven fabric produces less linting and is suitable for direct wound contact They are sterile to avoid contaminating the wound
Catalog No. 50-304-5503 Supplier Med Vet International Supplier No. 482272CS
Nonwoven Sponge McKesson Nonwoven / Rayon / Cellulose 4 X 4 Inch Square Sterile Nonwoven, 4 x 4 inch sterile sponges are ideal for moderate to high exuding wounds They have a rayon and cellulose absorbent core The nonwoven fabric produces less linting and is suitable for direct wound contact They are sterile to avoid contaminating the wound