Learn More
Med Vet International McKesson Sterile Nonwoven Sponge, 4 x 4 Inch, 16-2436, 2/Pack
Catalog No. 503045504
Encompass_Preferred
Nonwoven Sponge McKesson Nonwoven / Rayon / Cellulose 4 X 4 Inch Square Sterile Nonwoven, 4 x 4 inch sterile sponges are ideal for moderate to high exuding wounds They have a rayon and cellulose absorbent core The nonwoven fabric produces less linting and is suitable for direct wound contact They are sterile to avoid contaminating the wound
Catalog No. 50-304-5504 Supplier Med Vet International Supplier No. 482272PK
Nonwoven Sponge McKesson Nonwoven / Rayon / Cellulose 4 X 4 Inch Square Sterile Nonwoven, 4 x 4 inch sterile sponges are ideal for moderate to high exuding wounds They have a rayon and cellulose absorbent core The nonwoven fabric produces less linting and is suitable for direct wound contact They are sterile to avoid contaminating the wound