Learn More
Med Vet International I.V. Sponge, Medline, 2 x 2 Inch, 6-Ply, Sterile, NON256001, 70/Box
Catalog No. 503043175
Encompass_Preferred
Unique fenestration makes application easier, resists puckering, and enables the sponge to lie flat when wrapped around a drainage tube 6-ply rayon/polyester blend is highly absorbent, low-linting and fast wicking Precut to save time and eliminate the need for sterile scissors Drain not included
Catalog No. 50-304-3175 Supplier Med Vet International Supplier No. 1198567BX
Unique fenestration makes application easier, resists puckering, and enables the sponge to lie flat when wrapped around a drainage tube 6-ply rayon/polyester blend is highly absorbent, low-linting and fast wicking Precut to save time and eliminate the need for sterile scissors Drain not included