Learn More
Med Vet International McKesson NonSterile USP Type VII Gauze Sponge, 12-Ply, 2 x 2 Inch, 42222000, 200/Pack
Catalog No. 503043848
Encompass_Preferred
USP Type VII Gauze Sponge McKesson Cotton 12-Ply 2 X 2 Inch Square NonSterile Premium woven gauze sponges are a great first-aid must-have These USP Type VII gauze sponges consist of 100% cotton for comfort These woven cotton gauze pads have a 12-ply construction that makes them strong and highly absorbent 2 x 2 inches
Catalog No. 50-304-3848 Supplier Med Vet International Supplier No. 1043628PK
USP Type VII Gauze Sponge McKesson Cotton 12-Ply 2 X 2 Inch Square NonSterile Premium woven gauze sponges are a great first-aid must-have These USP Type VII gauze sponges consist of 100% cotton for comfort These woven cotton gauze pads have a 12-ply construction that makes them strong and highly absorbent 2 x 2 inches