Learn More
Med Vet International McKesson NonSterile USP Type VII Gauze Sponge, 12-Ply, 4 x 4 Inch, 42422000, 200/Pack
Catalog No. 503044008
Encompass_Preferred
USP Type VII Gauze Sponge McKesson Cotton 12-Ply 4 X 4 Inch Square NonSterile Ideal for wound dressings, wound packing and general wound care These USP Type VII gauze sponges consist of 100% cotton for comfort Woven cotton gauze pads have a 12-ply construction that makes them strong and highly absorbent 4 x 4 inches
Catalog No. 50-304-4008 Supplier Med Vet International Supplier No. 1043630PK
USP Type VII Gauze Sponge McKesson Cotton 12-Ply 4 X 4 Inch Square NonSterile Ideal for wound dressings, wound packing and general wound care These USP Type VII gauze sponges consist of 100% cotton for comfort Woven cotton gauze pads have a 12-ply construction that makes them strong and highly absorbent 4 x 4 inches