Learn More
Med Vet International McKesson Sterile Gauze Sponge, 8-Ply, 4 x 4 Inch, 16-4248, 1200/Case
Catalog No. 503044328
Encompass_Preferred
Gauze Sponge McKesson Cotton 8-Ply 4 X 4 Inch Square Sterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Sterile gauze sponge offers reduced linting properties Intended for single-use applications Peelable pouch simplifies access 4 x 4 inches
Catalog No. 50-304-4328 Supplier Med Vet International Supplier No. 762704CS
Gauze Sponge McKesson Cotton 8-Ply 4 X 4 Inch Square Sterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Sterile gauze sponge offers reduced linting properties Intended for single-use applications Peelable pouch simplifies access 4 x 4 inches