Learn More
Med Vet International McKesson NonSterile Gauze Sponge, 8-Ply, 3 x 3 Inch, 33082000, 200/Pack
Catalog No. 503044841
Encompass_Preferred
Gauze Sponge McKesson Cotton 8-Ply 3 X 3 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications 3 x 3 inches
Catalog No. 50-304-4841 Supplier Med Vet International Supplier No. 446030PK
Gauze Sponge McKesson Cotton 8-Ply 3 X 3 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications 3 x 3 inches