Learn More
Med Vet International Dynarex Sterile Tracheostomy Gauze Sponges, 100% Cotton, 4" X 4", 12 Ply, 300/Case
Catalog No. 502831194
Encompass_Preferred
Dynarex Resp-O2™ Tracheostomy Drain Sponges are premium-quality sterile dressings designed with a convenient pre-cut T-shaped notch for a comfortable fit around drains, catheters, chest tubes, IVs, and trach tubes. These reliable, highly absorbent pads are designed to help keep patients dry and comfortable. Pre-cut notch saves time and eliminates need for scissors Packed in peel-down pouches for sanitary handling Pouches come in open trays for easy access Sterile, not made with natural rubber latex
Catalog No. 50-283-1194 Supplier Med Vet International Supplier No. 35051
Dynarex Resp-O2™ Tracheostomy Drain Sponges are premium-quality sterile dressings designed with a convenient pre-cut T-shaped notch for a comfortable fit around drains, catheters, chest tubes, IVs, and trach tubes. These reliable, highly absorbent pads are designed to help keep patients dry and comfortable. Pre-cut notch saves time and eliminates need for scissors Packed in peel-down pouches for sanitary handling Pouches come in open trays for easy access Sterile, not made with natural rubber latex