Learn More
Med Vet International McKesson NonSterile Gauze Sponge, 12-Ply, 4 x 4 Inch, 12442000, 200/Pack
Catalog No. 503045505
Encompass_Preferred
Gauze Sponge McKesson Woven Cotton 12-Ply 4 X 4 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications H-fold design 4 x 4 inches
Catalog No. 50-304-5505 Supplier Med Vet International Supplier No. 446042PK
Gauze Sponge McKesson Woven Cotton 12-Ply 4 X 4 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications H-fold design 4 x 4 inches