Learn More
Med Vet International McKesson NonSterile Gauze Sponge, 16-Ply, 4 x 4 Inch, 16442000, 200/Pack
Catalog No. 503045487
Encompass_Preferred
Gauze Sponge McKesson Woven Cotton 16-Ply 4 X 4 Inch Square NonSterile 16 ply Nonsterile 100% cotton Appropriate for wound dressing, cleaning, prepping or packing Reduced linting 4 x 4 inches
Catalog No. 50-304-5487 Supplier Med Vet International Supplier No. 446043PK
Gauze Sponge McKesson Woven Cotton 16-Ply 4 X 4 Inch Square NonSterile 16 ply Nonsterile 100% cotton Appropriate for wound dressing, cleaning, prepping or packing Reduced linting 4 x 4 inches