Learn More
Med Vet International Gauze Sponge Curity™ 4 X 4 Inch, Sterile, 8-Ply Square, 2187-, 2/Pack, 25 Pack
Catalog No. 503044216
Encompass_Preferred
Eight ply USP Type VII: USP means gauze sponge meets United States Pharmacopeial Convention standards Packed in sterile pairs 10.2 x 10.2 centimeters
Catalog No. 50-304-4216 Supplier Med Vet International Supplier No. 688659TR
Eight ply USP Type VII: USP means gauze sponge meets United States Pharmacopeial Convention standards Packed in sterile pairs 10.2 x 10.2 centimeters