Learn More
Med Vet International McKesson NonSterile Gauze Sponge, 8-Ply, 2 x 2 Inch, 22802000, 5000/Case
Catalog No. 503044808
Encompass_Preferred
Gauze Sponge McKesson Woven Cotton 8-Ply 2 X 2 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications H-fold design 2 x 2 inches
Catalog No. 50-304-4808 Supplier Med Vet International Supplier No. 446037CS
Gauze Sponge McKesson Woven Cotton 8-Ply 2 X 2 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications H-fold design 2 x 2 inches