Learn More
Med Vet International McKesson NonSterile Gauze Sponge, 8-Ply, 4 x 4 Inch, 44802000, 200/Pack
Catalog No. 503045491
Encompass_Preferred
Gauze Sponge McKesson Woven Cotton 8-Ply 4 X 4 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications H-fold design 4 x 4 inches
Catalog No. 50-304-5491 Supplier Med Vet International Supplier No. 446041PK
Gauze Sponge McKesson Woven Cotton 8-Ply 4 X 4 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications H-fold design 4 x 4 inches