Learn More
Med Vet International Nonwoven Sponge Curity™ 2 X 2 Inch, NonSterile 4-Ply Square, 9022, 200/Bag
Catalog No. 503044758
Encompass_Preferred
Four ply Nonwoven Rayon/polyester blend construction Cost-effective Highly absorbent Virtually lint-free 2 x 2 inches
Catalog No. 50-304-4758 Supplier Med Vet International Supplier No. 151783BG
Four ply Nonwoven Rayon/polyester blend construction Cost-effective Highly absorbent Virtually lint-free 2 x 2 inches