Learn More
Med Vet International Nonwoven Sponge Curity™ 3 X 3 Inch, NonSterile 4-Ply Square, 9023, 200/Bag
Catalog No. 503043369
Encompass_Preferred
4 ply Nonwoven Rayon/polyester blend construction Cost-effective Highly absorbent Virtually lint-free 3 x 3 inches
Catalog No. 50-304-3369 Supplier Med Vet International Supplier No. 153822BG
4 ply Nonwoven Rayon/polyester blend construction Cost-effective Highly absorbent Virtually lint-free 3 x 3 inches