Learn More
Med Vet International Gauze Sponge Dermacea™ 2 X 2 Inch, Sterile 8-Ply Square, 441204, 3000/Case
Catalog No. 503044632
Encompass_Preferred
The Dermacea USP type VII gauze sponges are made of cotton for absorbency The material promotes wound drainage to help with healing The cotton gauze dressings have an eight-ply layer that offers protection The soft sponges are ideal for other applications such as wound prepping, packing, debriding and cleaning The sterile 2 x 2 inch gauze sponges won't infect wounds
Catalog No. 50-304-4632 Supplier Med Vet International Supplier No. 531576CS
The Dermacea USP type VII gauze sponges are made of cotton for absorbency The material promotes wound drainage to help with healing The cotton gauze dressings have an eight-ply layer that offers protection The soft sponges are ideal for other applications such as wound prepping, packing, debriding and cleaning The sterile 2 x 2 inch gauze sponges won't infect wounds