Learn More
Med Vet International Jorvet Laparotomy Sponges, Sterile, J0490BS, 4in x 18in, 2 Count
Catalog No. 5000495364
Encompass_Preferred
Prewashed 8-ply lap sponges,radio-opaque with loops. Useful in retaining bodystructures during deep abdominal surgery.
Catalog No. 50-004-95364 Supplier Med Vet International Supplier No. JOR490BS
Prewashed 8-ply lap sponges,radio-opaque with loops. Useful in retaining bodystructures during deep abdominal surgery.