Learn More
Med Vet International Jorvet Non-Woven Sponge Gauze, 4" x 4", 4-ply, 200/Pack, J0570C, 20 Packs/Case
Catalog No. 5000495304
Encompass_Preferred
Synthetic rayon and polyester non-woven fabric. These sponges are lint free, superior in strength and 50 percent more absorbable than cotton sponges.
Catalog No. 50-004-95304 Supplier Med Vet International Supplier No. JOR570C
Synthetic rayon and polyester non-woven fabric. These sponges are lint free, superior in strength and 50 percent more absorbable than cotton sponges.