Learn More
Med Vet International McKesson NonSterile Nonwoven Sponge, 4-Ply, 4 x 4 Inch, 94442000, 200/Pack
Catalog No. 503044807
Encompass_Preferred
Nonwoven Sponge McKesson Polyester / Rayon 4-Ply 4 X 4 Inch Square NonSterile McKesson nonwoven sponges have a blend of polyester and rayon, which is an alternative to cotton These 4-ply sponges are suited for general cleaning Each 4-inch x 4-inch nonwoven sponge is highly absorbent They are virtually lint-free McKesson nonwoven sponges are ideal for dressing, prepping and cleaning wounds Each pack contains 200 nonwoven sponges
Catalog No. 50-304-4807 Supplier Med Vet International Supplier No. 446036PK
Nonwoven Sponge McKesson Polyester / Rayon 4-Ply 4 X 4 Inch Square NonSterile McKesson nonwoven sponges have a blend of polyester and rayon, which is an alternative to cotton These 4-ply sponges are suited for general cleaning Each 4-inch x 4-inch nonwoven sponge is highly absorbent They are virtually lint-free McKesson nonwoven sponges are ideal for dressing, prepping and cleaning wounds Each pack contains 200 nonwoven sponges