Learn More
Med Vet International McKesson NonSterile Nonwoven Sponge, 4-Ply, 2 x 2 Inch, 16-607613, 200/Pack
Catalog No. 503045457
Encompass_Preferred
Nonwoven Sponge McKesson Cotton 4-Ply 2 X 2 Inch Square NonSterile This nonwoven 2 x 2 inch gauze is made with 100% cotton for softness and comfort The high absorbency helps keep the skin area surrounding wounds dry, clean and healthy The McKesson nonwoven sponge won't fray or produce lint over time A 4-ply design increases support, protection and absorbency
Catalog No. 50-304-5457 Supplier Med Vet International Supplier No. 481051PK
Nonwoven Sponge McKesson Cotton 4-Ply 2 X 2 Inch Square NonSterile This nonwoven 2 x 2 inch gauze is made with 100% cotton for softness and comfort The high absorbency helps keep the skin area surrounding wounds dry, clean and healthy The McKesson nonwoven sponge won't fray or produce lint over time A 4-ply design increases support, protection and absorbency