Learn More
Med Vet International McKesson Sterile Nonwoven Sponge, 6-Ply, 4 x 4 Inch, 16-4246, 2/Pack
Catalog No. 503043565
Encompass_Preferred
Nonwoven Sponge McKesson Polyester / Rayon 6-Ply 4 X 4 Inch Square Sterile McKesson nonwoven sponges are ideal for wound cleaning and dressing purposes Polyester-rayon blend absorbs moisture without linting Poly rayon blend has greater wicking abilities than cotton gauze Nonwoven sponges are strong and durable and don't tear easily 4 x 4 inches
Catalog No. 50-304-3565 Supplier Med Vet International Supplier No. 466523PK
Nonwoven Sponge McKesson Polyester / Rayon 6-Ply 4 X 4 Inch Square Sterile McKesson nonwoven sponges are ideal for wound cleaning and dressing purposes Polyester-rayon blend absorbs moisture without linting Poly rayon blend has greater wicking abilities than cotton gauze Nonwoven sponges are strong and durable and don't tear easily 4 x 4 inches