Learn More
Med Vet International Drain Sponge DuSoft 4 X 4 Inch Sterile 6-Ply, 84916, 2/Pack
Catalog No. 503043255
Encompass_Preferred
Provides superior absorption and are low-linting, fast-wicking, and non-adherent to wound beds 6 ply, nonwoven poly/rayon blend 4 x 4 inches
Catalog No. 50-304-3255 Supplier Med Vet International Supplier No. 645798PK
Provides superior absorption and are low-linting, fast-wicking, and non-adherent to wound beds 6 ply, nonwoven poly/rayon blend 4 x 4 inches