Learn More
Med Vet International McKesson Sterile Fluff Dressing, 6 x 6-3/4 Inch, 16-4265, 10/Box
Catalog No. 503045498
Encompass_Preferred
Fluff Dressing McKesson Cotton 6 X 6-3/4 Inch Rectangle Sterile 100% cotton Fast-wicking and highly absorbent Diagonal fold Conformable Packed in a soft tray Sterile 6 x 6-3/4 inches
Catalog No. 50-304-5498 Supplier Med Vet International Supplier No. 473032BX
Fluff Dressing McKesson Cotton 6 X 6-3/4 Inch Rectangle Sterile 100% cotton Fast-wicking and highly absorbent Diagonal fold Conformable Packed in a soft tray Sterile 6 x 6-3/4 inches