Learn More
Med Vet International McKesson Sterile Fluff Dressing, 6 x 6-3/4 Inch, 16-42626, 20/Box
Catalog No. 503044854
Encompass_Preferred
Fluff Dressing McKesson Cotton 6 X 6-3/4 Inch Rectangle Sterile Pre-washed 100% woven gauze Lofty and bulky for added cushioning and wound protection Fast-wicking and absorbent Use as a primary or secondary dressing Diagonal fold Conformable Packed in sterile 2s 6 x 6-3/4 inches
Catalog No. 50-304-4854 Supplier Med Vet International Supplier No. 446053BX
Fluff Dressing McKesson Cotton 6 X 6-3/4 Inch Rectangle Sterile Pre-washed 100% woven gauze Lofty and bulky for added cushioning and wound protection Fast-wicking and absorbent Use as a primary or secondary dressing Diagonal fold Conformable Packed in sterile 2s 6 x 6-3/4 inches