Learn More
Med Vet International McKesson Sterile x-Ray Detectable Gauze Sponge, 16-Ply, 4 x 4 Inch, 16-42446, 10/Tray
Catalog No. 503043160
Encompass_Preferred
X-Ray Detectable Gauze Sponge McKesson Cotton 16-Ply 4 X 4 Inch Square Sterile 16 ply Radiopaque Contains blue X-ray detectable barium sulfate strands for easy identification 4 x 4 inches
Catalog No. 50-304-3160 Supplier Med Vet International Supplier No. 472514TR
X-Ray Detectable Gauze Sponge McKesson Cotton 16-Ply 4 X 4 Inch Square Sterile 16 ply Radiopaque Contains blue X-ray detectable barium sulfate strands for easy identification 4 x 4 inches