Learn More
Med Vet International McKesson Sterile x-Ray Detectable Gauze Sponge, 12-Ply, 4 x 8 Inch, 16-42846, 10/Tray
Catalog No. 503043162
Encompass_Preferred
X-Ray Detectable Gauze Sponge McKesson Cotton 12-Ply 4 X 8 Inch Rectangle Sterile 12 ply Radiopaque Contains blue X-ray detectable barium sulfate strands for easy identification 4 x 8 inches
Catalog No. 50-304-3162 Supplier Med Vet International Supplier No. 472515TR
X-Ray Detectable Gauze Sponge McKesson Cotton 12-Ply 4 X 8 Inch Rectangle Sterile 12 ply Radiopaque Contains blue X-ray detectable barium sulfate strands for easy identification 4 x 8 inches