Learn More
Med Vet International Gauze Sponge Curity™ 4 X 4 Inch, Sterile 16-Ply Square, 7605-, 1280/Case
Catalog No. 503044795
Encompass_Preferred
16 ply USP Type VII: USP means gauze sponge meets United States Pharmacopeial Convention standards Conveniently indexed in 10s for faster, easier handling 4 x 4 inches
Catalog No. 50-304-4795 Supplier Med Vet International Supplier No. 1158693CS
16 ply USP Type VII: USP means gauze sponge meets United States Pharmacopeial Convention standards Conveniently indexed in 10s for faster, easier handling 4 x 4 inches