Learn More
Med Vet International Jorvet Laparotomy Sponges, Non-Sterile, J0490A, 18in x 18in, 5 Count
Catalog No. 5000495292
Encompass_Preferred
Prewashed 8-ply lap sponges, radio-opaque with loops. Useful in retaining body structures during deep abdominal surgery.
Catalog No. 50-004-95292 Supplier Med Vet International Supplier No. JOR490A
Prewashed 8-ply lap sponges, radio-opaque with loops. Useful in retaining body structures during deep abdominal surgery.