Learn More
Med Vet International Jorvet Laparotomy Sponges, Non-Sterile, J0490B, 4in x 18in, 5 Count
Catalog No. 5000495156
Encompass_Preferred
Prewashed 8-ply lap sponges,radio-opaque with loops. Useful in retaining bodystructures during deep abdominal surgery.
Catalog No. 50-004-95156 Supplier Med Vet International Supplier No. JOR490B
Prewashed 8-ply lap sponges,radio-opaque with loops. Useful in retaining bodystructures during deep abdominal surgery.