Learn More
Aviva Systems Biology Corp SPNS1 Antibody - N-terminal region (ARP50029_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50029_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPNS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SPNS1. Alternate names: FLJ38358, HSpin1, LAT, PP2030, SPIN1, SPINL, nrsApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60247