Learn More
Aviva Systems Biology Corp SPOCK2 Antibody - N-terminal region (ARP62236_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62236_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPOCK2. It was validated on Western Blot. . Alternate names: FLJ97039, testican-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-625