Learn More
Aviva Systems Biology Corp SPOCK3 Antibody - middle region (ARP44921_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44921_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPOCK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SPOCK3. Alternate names: HSAJ1454, TES-3, TICN3ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58937