Learn More
ABclonal Technology SPOCK3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1284920UL
Encompass_Preferred
This gene encodes a member of a novel family of calcium-binding proteoglycan proteins that contain thyroglobulin type-1 and Kazal-like domains. The encoded protein and may play a role in adult T-cell leukemia by inhibiting the activity of membrane-type matrix metalloproteinases. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Catalog No. 50-001-16030