Learn More
Aviva Systems Biology Corp SPON2 Antibody - N-terminal region (ARP52335_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52335_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPON2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SPON2. Alternate names: DIL-1, DIL1, M-spondin, Mindin, MINDIN, M-SPONDINApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-60715