Learn More
Keystone Safety Sponbond Polypropylene Hood with Elastic Closure
SDP
Catalog No. 01257801
Change view
Click to view available options
Size:
Large
Catalog No. Size
01-257-801 Large
1 options
Catalog No. 01-257-801 Supplier Keystone Safety Supplier No. HNWIECROWNBLUE
Add to Cart
Add to Cart
Made from breathable polypropylene with a secure elastic closure, it delivers comfortable protection while helping reduce contamination risks.
  • Head and hair coverage for cleanrooms, laboratories, and controlled environments
  • Lightweight, breathable polypropylene material
  • Elastic closure for a secure fit
  • Helps reduce contamination risks from hair and particulate matter
  • Low-lint construction suitable for controlled environments
  • 100% latex-free
  • Disposable, single-use design
  • Standard size
Specifications
Product Type Polypropylene Hood
Size Large
Material Spunbond Polypropylene
Color Blue
Closure Type Elastic
Disposable Single-use
Cleanroom Class Class 10,000 (ISO 7)