Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Spondin-2/SPON2 Protein, Human (HEK293, His) | ≥ 95% | Approximately 38-47 kDa | 250 UG
SDP

Catalog No. 5000399919
Encompass_Preferred

Spondin-2/SPON2 Protein, Human (HEK293, His) | ≥ 95% | Approximately 38-47 kDa | 250 UG

Catalog No. 50-003-99919 Supplier Medchemexpress LLC Supplier No. 250UGHYP70142
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spondin-2/SPON2 Protein, Human (HEK293, His) | ≥ 95% | Approximately 38-47 kDa | 250 UG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.