Learn More
Andwin Scientific SPONGE 5 1/2 X 3 5/8 X 1 12/PK
SDP
Catalog No. NC9822663
Encompass_Preferred
The Fine-Pore Cellulose Sponge features durability and convenience as it rapidly absorbs and cleans spills or liquids. This product greatly combats tearing and abrasion. It can hold up to 25 times their own weight in water. This product is sold in a
Catalog No. NC9822663 Supplier Andwin Scientific Supplier No. 58540047
The Fine-Pore Cellulose Sponge features durability and convenience as it rapidly absorbs and cleans spills or liquids. This product greatly combats tearing and abrasion. It can hold up to 25 times their own weight in water. This product is sold in a