Learn More
Med Vet International Non-Woven Avant Gauze Drain Sponges, 2X2, Sterile, 2/Pack (Box Of 35 Pk)
Catalog No. 502831101
Encompass_Preferred
Avant Gauze Drain Sponges: This sponge features an extra fenestration which allows it to lay flat when wrapped around a drainage tube. Without it, the sponge will pucker, decreasing effectiveness by allowing fluids to pool around the drain tube. It's a simple feature that most other drain sponges do not offer. Includes all the benefits of non-woven Avant Gauze. Each 6-ply rayon/polyester sponge is highly absorbent, low-linting and fast wicking to wound beds. 2" x 2", 6-ply, sterile 2's.
Catalog No. 50-283-1101 Supplier Med Vet International Supplier No. NON256001Z
Avant Gauze Drain Sponges: This sponge features an extra fenestration which allows it to lay flat when wrapped around a drainage tube. Without it, the sponge will pucker, decreasing effectiveness by allowing fluids to pool around the drain tube. It's a simple feature that most other drain sponges do not offer. Includes all the benefits of non-woven Avant Gauze. Each 6-ply rayon/polyester sponge is highly absorbent, low-linting and fast wicking to wound beds. 2" x 2", 6-ply, sterile 2's.