Learn More
Sklar Instruments Econo Foerster Sponge Forceps, Sklar, 9-1/2 in., Fenestrated, Straight, Serrated, Stainless Steel. PK/12.
SDP
Catalog No. 501437667 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Foerster Sponge Forceps, Sklar, 9-1/2 in., Fenestrated, Straight, Serrated, Stainless Steel. Econo Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site and hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster Sponge Forceps that are longer in length are often used in bariatric surgeries and the curved version may be used for removing the placenta in OB/GYN procedures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with oval, fenestrated, serrated tips and a length of 9-1/2 inches. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7667 Supplier Sklar Instruments Supplier No. 21577
Econo Foerster Sponge Forceps, Sklar, 9-1/2 in., Fenestrated, Straight, Serrated, Stainless Steel. Econo Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site and hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster Sponge Forceps that are longer in length are often used in bariatric surgeries and the curved version may be used for removing the placenta in OB/GYN procedures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with oval, fenestrated, serrated tips and a length of 9-1/2 inches. They are sold in boxes of 12.