Learn More
Med Vet International McKesson Sterile Gauze Sponge, 12-Ply, 2 x 2 Inch, 16-4221, 2400/Case
Catalog No. 503045017
Encompass_Preferred
Gauze Sponge McKesson Cotton 12-Ply 2 X 2 Inch Square Sterile 12 ply Sterile 100% cotton Appropriate for wound dressing, cleaning, prepping or packing Reduced linting Packed in convenient, peelable pouch 2 x 2 inches
Catalog No. 50-304-5017 Supplier Med Vet International Supplier No. 501657CS
Gauze Sponge McKesson Cotton 12-Ply 2 X 2 Inch Square Sterile 12 ply Sterile 100% cotton Appropriate for wound dressing, cleaning, prepping or packing Reduced linting Packed in convenient, peelable pouch 2 x 2 inches