Learn More
Med Vet International McKesson NonSterile Gauze Sponge, 12-Ply, 2 x 2 Inch, 22122000, 8000/Case
Catalog No. 503045460
Encompass_Preferred
Gauze Sponge McKesson Cotton 12-Ply 2 X 2 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties 2 x 2 inches
Catalog No. 50-304-5460 Supplier Med Vet International Supplier No. 446029CS
Gauze Sponge McKesson Cotton 12-Ply 2 X 2 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties 2 x 2 inches