Learn More
Med Vet International Gauze Sponge Curity™ 2 X 2 Inch, NonSterile 12-Ply Square, 2252, 8000/Case
Catalog No. 503045823
Encompass_Preferred
12 ply USP Type VII USP means gauze sponge meets United States Pharmacopeial Convention standards 2 x 2 inches
Catalog No. 50-304-5823 Supplier Med Vet International Supplier No. 9979CS
12 ply USP Type VII USP means gauze sponge meets United States Pharmacopeial Convention standards 2 x 2 inches