Learn More
Med Vet International McKesson NonSterile Gauze Sponge, 12-Ply, 3 x 3 Inch, 33122000, 200/Box
Catalog No. 503043450
Encompass_Preferred
Gauze Sponge McKesson Cotton 12-Ply 3 X 3 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications 3 x 3 inches
Catalog No. 50-304-3450 Supplier Med Vet International Supplier No. 373776BX
Gauze Sponge McKesson Cotton 12-Ply 3 X 3 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications 3 x 3 inches