Learn More
Med Vet International Gauze Sponge Dermacea™ 4 X 4 Inch, Sterile 12-Ply Square, 442214, 10/Pack
Catalog No. 503044602
Encompass_Preferred
These Dermacea gauze sponges are made of cotton for absorbency They meet USP Type VII gauze requirements These gauze sponges have a twelve-ply layer that offers protection They're ideal for other applications such as wound prepping, packing, debriding and cleaning They're sterile to prevent infecting wounds 4 x 4 inches
Catalog No. 50-304-4602 Supplier Med Vet International Supplier No. 564971PK
These Dermacea gauze sponges are made of cotton for absorbency They meet USP Type VII gauze requirements These gauze sponges have a twelve-ply layer that offers protection They're ideal for other applications such as wound prepping, packing, debriding and cleaning They're sterile to prevent infecting wounds 4 x 4 inches