Learn More
Med Vet International Gauze Sponge Curity™ 4 X 4 Inch, NonSterile 12-Ply Square, 2634-, 200/Bag
Catalog No. 503043844
Encompass_Preferred
Twelve ply Conveniently indexed in 10s for faster, easier handling USP Type VII: USP means gauze sponge meets United States Pharmacopeial Convention standards 4 x 4 inches
Catalog No. 50-304-3844 Supplier Med Vet International Supplier No. 10001BG
Twelve ply Conveniently indexed in 10s for faster, easier handling USP Type VII: USP means gauze sponge meets United States Pharmacopeial Convention standards 4 x 4 inches