Learn More
Med Vet International Gauze Sponge Curity™ 4 X 4 Inch, NonSterile 12-Ply Square, 2634-, 2000/Case
Catalog No. 503043845
Encompass_Preferred
Twelve ply Conveniently indexed in 10s for faster, easier handling USP Type VII: USP means gauze sponge meets United States Pharmacopeial Convention standards 4 x 4 inches
Catalog No. 50-304-3845 Supplier Med Vet International Supplier No. 10001CS
Twelve ply Conveniently indexed in 10s for faster, easier handling USP Type VII: USP means gauze sponge meets United States Pharmacopeial Convention standards 4 x 4 inches